Anti-CCNC Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-226
PRSI63-226EA 620 EUR
PRSI63-226
Anti-CCNC Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000.

Type: Primary
Antigen: CCNC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 221-249
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR