Anti-VARS Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-235
PRSI63-235EA 620 EUR
PRSI63-235
Anti-VARS Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50.

Type: Primary
Antigen: VARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 251-279
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR