Anti-LRPAP1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-504
PRSI63-504EA 660 EUR
PRSI63-504
Anti-LRPAP1 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For IHC-P starting dilution is: 1:10~50. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: LRPAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 281-309
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR