Anti-AGPAT4 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-601
PRSI63-601EA 620 EUR
PRSI63-601
Anti-AGPAT4 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: AGPAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 176-202
Host: Rabbit
Isotype: Ig
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR