Anti-ACY3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
63-608
PRSI63-608EA 620 EUR
PRSI63-608
Anti-ACY3 Rabbit Polyclonal Antibody
Antikörper
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.

Type: Primary
Antigen: ACY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 66-95
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR