Anti-RASSF6 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
6543
PRSI6543EA 435 EUR
PRSI6543
Anti-RASSF6 Rabbit Polyclonal Antibody
Antikörper
RASSF6 Antibody: RASSF family proteins are tumor suppressors that are frequently downregulated during the development of human cancer. RASSF6 (Ras association (RalGDS/AF-6) domain family member 6), a member of RASSF family, acts as a Ras effector protein / tumor suppressor. RASSF6 contains one Ras-associating domain and one SARAH domain. It is involved in the induction of apoptosis, through both caspase-dependent and caspase-independent pathways. Defects in RASSF6 may facilitate viral NF kappa -B pathway and may play a vital role in infection.

Type:
Antigen: RASSF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR