Anti-SPATA3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
6551
PRSI6551EA 435 EUR
PRSI6551
Anti-SPATA3 Rabbit Polyclonal Antibody
Antikörper
SPATA3 Antibody: The testis and spermatogenesis cell apoptosis related gene 1 (TSARG1) protein, also known as SPATA3, is highly expressed in testis, and to a lower extent in other tissues. SPATA3 contains a highly conserved J domain, which is present in all DnaJ-like proteins, and is thus a novel member of the HSP40 protein family. SPATA3 is thought to be involved in apoptosis of spermatogenic cells.

Type:
Antigen: SPATA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR