Anti-Thymopoietin Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
6605
PRSI6605EA 435 EUR
PRSI6605
Anti-Thymopoietin Rabbit Polyclonal Antibody
Antikörper
Thymopoietin Antibody: Thymopoietin (TMPO), also known as LAP2, is a widely expressed lem-domain-containing nuclear protein that is thought to play an important role in the regulation of nuclear architecture by binding lamin B1 and chromosomes in a manner regulated by phosphorylation during mitosis. Thymopoietin associates with nucleoplasmic lamins A/C, which are encoded by the dilated cardiomyopathy (DCM) gene LMNA; mutations in Thymopoietin disrupt this binding, suggesting that Thymopoietin may be a candidate disease gene for DCM.

Type:
Antigen: Thymopoietin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR