Anti-FLT3 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
7027
PRSI7027EA 435 EUR
PRSI7027
Anti-FLT3 Rabbit Polyclonal Antibody
Antikörper
FLT3 Antibody: FLT3 is a member of the class III receptor tyrosine kinase family and its internal tandem duplication (ITD) is preferentially seen in acute myeloid leukemia and myelodysplastic syndrome among various hematological malignancies. It is a membrane bound glycosylated protein that is expressed primarily in early hematopoietic progenitor cells. Upon binding its ligand FL, FLT3 activates multiple signaling pathways including the PI3K/Akt and Ras/MEK/MAPK pathways. In cells expressing FLT3-ITD, these pathways are constitutively on, resulting in abnormal cell growth and anti-apoptotic activity.

Type:
Antigen: FLT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR