Anti-CARM1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
7219
PRSI7219EA 435 EUR
PRSI7219
Anti-CARM1 Rabbit Polyclonal Antibody
Antikörper
CARM1 Antibody: Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in signal transduction, metabolism of nascent pre-RNA, and transcriptional activation. CARM1 functions as a transcriptional co-activator for various nuclear receptors and NF-κB. It has also been shown to methylate histone H3 arginines, inhibiting the binding of corepressors and protecting chromatin from deacetylation, thereby facilitating transcription.

CARM1 Antibody is affinity chromatography purified via peptide column.

CARM1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CARM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR