GRIP1 Antibody: Glutamate receptors play an important role in neural plasticity, development and degeneration. The glutamate receptor interacting proteins, GRIP1 and GRIP2, members of the PDZ domain-containing protein family, mediate the trafficking and membrane organization of a number of transmembrane proteins. GRIP1 and GRIP2 specifically bind to the AMPA receptor subunits, GluR 2/3 and are involved in the targeting of GluR 2/3 to the synapse. GRIP1 is expressed in early development before the expression of AMPA receptors, while GRIP2 expression parallels that of AMPA receptors during later developmental stages. GRIP1 and GRIP2 may be involved in the induction of cerebellar long-term depression (LTD).
GRIP1 Antibody is affinity chromatography purified via peptide column.
GRIP1 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GRIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat