Anti-FOXA1 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
7255
PRSI7255EA 435 EUR
PRSI7255
Anti-FOXA1 Rabbit Polyclonal Antibody
Antikörper
FOXA1 Antibody: FOXA1 is one of three members of the FOXA family, a subset of the forkhead family of transcription factors which play vital roles in development. It has also been implicated in the development of a number of other organs including the androgen and estrogen regulated tissues of the breast and prostate. FOXA1 modulates the transcriptional activity of nuclear hormone receptors and regulates apoptosis by inhibiting the expression of BCL-2. It is an essential protein for the transcriptional activity of both androgen receptor (AR) and estrogen receptor-beta (ER). FOXA1 plays a pivotal role from early stage cancer through to drug resistant and metastatic disease. FOXA1 is not only an attractive therapeutic target but could potentially function as a novel biomarker.

FOXA1 Antibody is affinity chromatography purified via peptide column.

FOXA1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR