Anti-FOXA2 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
7257
PRSI7257EA 435 EUR
PRSI7257
Anti-FOXA2 Rabbit Polyclonal Antibody
Antikörper
FOXA2 Antibody: FOXA2 is one of three members of the FOXA family, a subset of the forkhead family of transcription factors which play vital roles in development. FOXA2 was initially identified through library screening as a closely related homolog of FOXA1. Both FOXA2 and FOXA1 act as transcriptional activators in adult liver and also play a role in body axis formation, neural tube patterning and definitive endoderm formation during gastrulation.

FOXA2 Antibody is affinity chromatography purified via peptide column.

FOXA2 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR