Anti-GOLGA5 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
7387
PRSI7387EA 435 EUR
PRSI7387
Anti-GOLGA5 Rabbit Polyclonal Antibody
Antikörper
GOLGA5 Antibody: GOLGA5 (Golgi autoantigen, subfamily A, member 5), also known as GOLGA84, RFG5 (RET-fused gene 5 protein) or GOLIM5, is a ubiquitously expressed single-pass type II coiled-coil membrane protein that is involved in maintaining Golgi structure. It is localized to the Golgi apparatus and predominantly found on membranes at the cis side of the Golgi stack (1,2). As a mitotic phosphoprotein, GOLGA5 binds to active Rab 1 and associates with CASP (an isoform of CDP) in a golgin-tethering complex that is believed to play a role in intra-Golgi retrograde transport. Defects in GOLGA5 are a cause of thyroid papillary carcinoma (TPC).

GOLGA5 Antibody is affinity chromatography purified via peptide column.

GOLGA5 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GOLGA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR