Anti-FCGR2A Mouse Monoclonal Antibody [clone: 6C4]

Lieferant: ProSci Inc.
76-193
PRSI76-193EA 377 EUR
PRSI76-193
Anti-FCGR2A Mouse Monoclonal Antibody [clone: 6C4]
Antikörper
The 6C4 monoclonal antibody specifically recognizes two isoforms of human CD32, Fc gamma RIIA and Fc gamma RIIB. From alternative mRNA splicing, 6 isoforms of CD32 have been reported. CD32 is a 40 kDa polymorphic transmembrane glycoprotein expressed on monocytes, macrophages, graulocytes, B cells, and platelets. The receptor has low affinity toward IgG and is involved in immunomodulation, phagocytosis, and the inflammation response. The 6C4 antibody in a rosette blocking assay is reported to inhibit Ig binding

Type: Primary
Antigen: FCGR2A
Clonality: Monoclonal
Clone: 6C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR