Anti-Il2ra Rat Monoclonal Antibody [clone: PC61.5] (APC (Allophycocyanin))

Lieferant: ProSci Inc.
76-520
PRSI76-520EA 271 EUR
PRSI76-520
Anti-Il2ra Rat Monoclonal Antibody [clone: PC61.5] (APC (Allophycocyanin))
Antikörper
The PC61.5 antibody specifically reacts with mouse CD25, the 55 kDa low-affinity Interleukin-2 Receptor alpha chain (IL-2R alpha), expressed on early progenitors of T and B lineage, and on B and T cells. Together with CD122 (IL-2 Receptor beta) and CD 132 (IL-2 Receptor gamma c, the common gamma chain), CD25 forms high-affinity receptor complexes for IL-2. Resting B and T cells and natural killer cells do not express IL-2R alpha. Cd25 is also expressed on the dendritic cells, and it enhances lymphocyte differentiation and activation.The PC61.5 antibody block the binding of IL-2 to both high-affinity and low-affinity receptors.

Type: Primary
Antigen: Il2ra
Clonality: Monoclonal
Clone: PC61.5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR