Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.
76-527
PRSI76-527EA 199 EUR
PRSI76-527
Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR