Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (APC (Allophycocyanin))

Lieferant: ProSci Inc.
76-533
PRSI76-533EA 409 EUR
PRSI76-533
Anti-Ptprc Rat Monoclonal Antibody [clone: 30-F11] (APC (Allophycocyanin))
Antikörper
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: 30-F11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR