Anti-Cd69 Hamster Monoclonal Antibody [clone: H1.2F3]

Lieferant: ProSci Inc.
76-817
PRSI76-817EA 409 EUR
PRSI76-817
Anti-Cd69 Hamster Monoclonal Antibody [clone: H1.2F3]
Antikörper
The H1.2F3 monoclonal antibody specifically reacts with human CD69, the 27 to 33 kDa type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: Cd69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR