Anti-HLA-DRA Mouse Monoclonal Antibody [clone: LN3] (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci Inc.
77-008
PRSI77-008EA 522 EUR
PRSI77-008
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: LN3] (FITC (Fluorescein Isothiocyanate))
Antikörper
The LN3 monoclonal antibody specifically reacts with the human leukocyte antigen-DR (HLA-DR), a MHC class II heterodimeric surface glycoprotein. HLA-DR is expressed on the antigen presenting surfaces of monocytes, macrophages, dendritic cells, activated T cells, and B cells. It is integral to peptide presentation to CD4+ T cells.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: LN3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR