Anti-Nucleocapsid Phosphoprotein Rabbit Polyclonal Antibody (Biotinylated)

Lieferant: ProSci Inc.
9099-BIOTIN
PRSI9099-BIOTINEA 547 EUR
PRSI9099-BIOTIN
Anti-Nucleocapsid Phosphoprotein Rabbit Polyclonal Antibody (Biotinylated)
Antikörper
SARS-CoV-2 (COVID-19) Nucleocapsid Antibody (biotin) can be stored at 4 °C for three months and −20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.

Type: Primary
Antigen: N
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR