Anti-Cd4 Rat Monoclonal Antibody [clone: GK1.5] (PE (Phycoerythrin)/Texas Red®)

Lieferant: ProSci Inc.
98-587
PRSI98-587EA 549 EUR
PRSI98-587
Anti-Cd4 Rat Monoclonal Antibody [clone: GK1.5] (PE (Phycoerythrin)/Texas Red®)
Antikörper
Type: Primary
Antigen: Cd4
Clonality: Monoclonal
Clone: [GK1.5]
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR