Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (PE (Phycoerythrin)/Texas Red®)

Lieferant: ProSci Inc.
98-617
PRSI98-617EA 620 EUR
PRSI98-617
Anti-Cd8a Rat Monoclonal Antibody [clone: 53-6.7] (PE (Phycoerythrin)/Texas Red®)
Antikörper
Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: [53-6.7]
Conjugation: PE (Phycoerythrin)/Texas Red®
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR