Anti-HAX1 Mouse Monoclonal Antibody [clone: 9F3D11]
Lieferant: ProSci Inc.
PM-4718
PRSIPM-4718EA 435 EUR
PRSIPM-4718
Anti-HAX1 Mouse Monoclonal Antibody [clone: 9F3D11]
Antikörper
Hax1a Monoclonal Antibody: The HS-1 associated protein X-1 (Hax1) was initially identified in a yeast two-hybrid assay on the basis of its ability to bind to the hemapoietic cell-specific protein 1 (HS-1). Hax1 possesses anti-apoptotic activity and is structurally related to Bcl-2 family members, including the presence of BH1- and BH2-like domains. It has recently been shown to interact with HIV viral protein R (Vpr), a protein required for viral pathogenesis of HIV and linked to T-cell apoptosis through activation of caspases 3 and 9. Other studies indicate that Hax1-mediated processing ofMore Product Information