Anti-HIV-1 p24 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: ProSci Inc.
PM-6335-HRP
PRSIPM-6335-HRPEA 435 EUR
PRSIPM-6335-HRP
Anti-HIV-1 p24 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
HIV-1 p24 Monoclonal Antibody: The human immunodeficiency virus type 1 (HIV-1) particle consists of an envelope, a core and the region between the two termed matrix. The HIV-1 Gag protein is a late structural protein that contains four proteins: matrix (p17), capsid (p24), nucleocapsid (p7) and the p6 protein. The p24 constitutes the major core component of the virus and shows high degree of sequence conservation among HIV isolates. The Gag p24 has been used as an integral part of multicomponent HIV-1 vaccines.
PM-6335-HRP can be used for detection of p24 by Western blot or ELISA at 0.2 – 0.5μg/mL.
HIV-1 p24 antibody is supplied as protein A purified IgG1.
HIV-1 p24 antibody is supplied in PBS containing 0.02% thimerosal.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: HIV-1 p24
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus