Anti-FGF1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: ProSci Inc.
XP-5136BT
PRSIXP-5136BTEA 509 EUR
PRSIXP-5136BT
Anti-FGF1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Anti-hFGF-acidic specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FGF1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR