Anti-CCL14 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
XP-5151
PRSIXP-5151EA 509 EUR
PRSIXP-5151
Anti-CCL14 Rabbit Polyclonal Antibody
Antikörper
Anti-hHCC-1 specific antibody was purified by affinity chromatography employing immobilized hHCC-1 matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCL14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR