Anti-IFNG Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
XP-5155
PRSIXP-5155EA 509 EUR
PRSIXP-5155
Anti-IFNG Rabbit Polyclonal Antibody
Antikörper
IFN gamma receptor beta is part of the receptor for interferon gamma. This class II cytokine receptor pairs with CDw119 to form the IFN gamma receptor and is an integral part of the IFN gamma signal transduction pathway. CDw119 serves as the IFN gamma binding chain and associates with the IFN gamma beta chain which is required for receptor signaling. The extracellular portion of both the IFN gamma receptor alpha and beta chains must be species matched. The IFN gamma receptor beta chain is expressed on T and B cells, NK cells, monocytes/ macrophages, and fibroblasts. Binding of IFN gamma induces receptor dimerization, internalization, Jak1 and Jak2 protein kinase activation and, ultimately, STAT1 activation. It is also likely to interact with GAF. IFN gamma initiates and regulates a variety of immune responses and is required for signal transduction. Contains 2 fibronectin type III domains. Defects in IFN gamma Receptor beta are a cause of mendelian susceptibility to mycobacterial disease (MSMD), a rare condition that confers predisposition to illness caused by several mycobacteria strains

Anti-hIFN gamma specific antibody was purified by affinity chromatography employing immobilized hIFN gamma matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR