Anti-IFNG Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
XP-5156
PRSIXP-5156EA 509 EUR
PRSIXP-5156
Anti-IFNG Rabbit Polyclonal Antibody
Antikörper
Anti-mIFN-gamma specific antibody was purified by affinity chromatography employing immobilized mIFN-gamma matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IFNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR