Anti-IL7 Rabbit Polyclonal Antibody

Lieferant: ProSci Inc.
XP-5197
PRSIXP-5197EA 509 EUR
PRSIXP-5197
Anti-IL7 Rabbit Polyclonal Antibody
Antikörper
Iterleukin 7 is a lymphoid cell growth factor that affects pre-B, pro-B, and early T cells. IL-7 was previously known as pre-B cell growth factor and lymphopoietin 1. IL-7 supports the growth of early B cells from long-term lymphoid bone marrow cultures. It is mitogenic to thymocytes and enhances the response of cells to other stimuli such as polyhydroxyalkanoate (PHA) and concanavalin A (ConA). IL-7 stimulates the proliferation of CD4+/CD8+ cells.

Anti-hIL-7 specific antibody was purified by affinity chromatography employing immobilized hIL-7 matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IL7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR