Anti-PF4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: ProSci Inc.
XP-5256BT
PRSIXP-5256BTEA 509 EUR
PRSIXP-5256BT
Anti-PF4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Platelet factor 4 (PF-4) is a 70 amino acid protein that is released from the alpha granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF-4 probably has a role in inflammation and wound repair.

Anti-hPF-4 specific antibody was purified by affinity chromatography and then biotinylated.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PF4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR