Anti-GSTO1 Chicken Polyclonal Antibody

Lieferant: ProSci Inc.
XW-7952
PRSIXW-7952EA 773 EUR
PRSIXW-7952
Anti-GSTO1 Chicken Polyclonal Antibody
Antikörper
FUNCTION: Exhibits glutathione-dependent thiol transferase and dehydroascorbate reductase activities.
CATALYTIC ACTIVITY: RX + glutathione = HX + R-S-glutathione.
SUBUNIT: Homodimer.
SIMILARITY: Belongs to the GST superfamily. Omega family.
SUBCELLULAR LOCATION: Cytoplasm.
TISSUE SPECIFICITY: Ubiquitous. Highest expression in liver, skeletal muscle and heart. Lowest expression in brain, placenta and lung.

Glutathione transferase omega-1 antibody can be used for the detection of Glutathione transferase omega-1 by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Phosphate-Buffered Saline. No preservatives added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GSTO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR