Anti-SLC12A2 Chicken Polyclonal Antibody
Lieferant: ProSci Inc.
XW-8138
PRSIXW-8138EA 826 EUR
PRSIXW-8138
Anti-SLC12A2 Chicken Polyclonal Antibody
Antikörper
FUNCTION: Electrically silent transporter system. Mediates sodium and chloride reabsorption. Plays a vital role in the regulation of ionic balance and cell volume.
SIMILARITY: Belongs to the SLC12A transporter family.
SUBCELLULAR LOCATION: Membrane; multi-pass membrane protein.
TISSUE SPECIFICITY: Expressed in many tissues.
Solute carrier family 12 member 2 antibody can be used for the detection of Solute carrier family 12 member 2 by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC12A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine, Chimpanzee