MSC Characterization Antibody Panel
Lieferant: STEMCELL Technologies
100-0354
STMC100-0354EA 779 EUR
STMC100-0354
MSC Characterization Antibody Panel
Antikörper
Multi-color flow cytometry kit for characterization of human mesenchymal stromal cells
  • Mesenchymal Stromal cells
The MSC Characterization Antibody Panel is designed for the phenotypic analysis of human mesenchymal stromal cells (MSCs). This kit contains three fluorochrome-conjugated antibodies for the identification of human MSCs, and a fluorochrome-conjugated antibody for the identification of non-MSCs. When measured by flow cytometry, ≥95% of the MSC population must express CD73, CD90, and CD105, and these cells must lack expression (≤2% positive) of CD45. These reagents provide single-step labeling for the verification of human MSCs.The MSC characterization antibody panel can be used to identify human MSCs derived from various tissues such as bone marrow MSCs, pluripotent stem cell-derived mesenchymal progenitor cells, adipose-derived MSCs, dental pulp-derived MSCs, and umbilical cord-derived MSCs.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').