Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (Biotin)

Lieferant: STEMCELL Technologies
60019BT 100-0425 100-0424
STMC60019BTEA 362 EUR
STMC60019BT STMC100-0425 STMC100-0424
Anti-CD45R (B220) Rat Monoclonal Antibody [clone: RA3-6B2] (Biotin)
Antikörper
Rat monoclonal IgG2a antibody against human, mouse, cat CD45R (B220), biotin-conjugated.

  • B cells
  • Leukocytes
  • Lymphocytes

The RA3-6B2 antibody reacts with an epitope on the extracellular domain of the exon A‐restricted isoform of mouse CD45. CD45 is a member of the protein tyrosine phosphatase family and functions in a number of immunoregulatory processes, including cell activation, growth, differentiation, and oncogenic transformation. The large cytoplasmic portion of CD45 contains two tyrosine phosphatase domains (one of which is enzymatically active) that are involved in modulating the function of Src kinases such as Lck and Fyn. Several isoforms of CD45 have been identified, the expression of which differs according to cell type and functional status. Alternative splicing of three exons encoding the extracellular RA, RB, and RC polypeptide sequences gives rise to up to 8 isoforms with molecular masses in the range of 200 - 220 kDa. The exon A‐restricted isoform of mouse CD45, termed CD45R, contains the RA, RB, and RC regions and is the largest of the isoforms. CD45R is expressed by the B cell lineage from early pro‐B to mature B cells, with its expression downregulated during differentiation to plasma cells. CD45R is also expressed on subsets of T, NK, dendritic, and lymphokine-activated killer (LAK) cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse B Cell Isolation Kit (Catalog #19854).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD45R (B220)
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Cat, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR