Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5] (PE)

Lieferant: STEMCELL Technologies
100-1605
STMC100-1605EA 58.8 EUR
STMC100-1605
Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5] (PE)
Antikörper
Rat monoclonal antibody against mouse, hamster CD4, PE-conjugated.

  • Dendritic cells
  • T cells
  • T cells CD4

This monoclonal antibody reacts with mouse cluster of differentiation 4 (CD4), a 55 kDa single chain type 1 transmembrane glycoprotein belonging to the immunoglobulin (Ig) superfamily. CD4 contains four extracellular Ig-like domains; D1 - D4. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells; T helper/inducer cells, and at lower levels on dendritic cells. Use this recombinant monoclonal antibody in applications requiring detection of CD4, such as flow cytometry (FC), immunohistochemistry (IHC), and immunofluorescence (IF). Unlike humans, CD4 is not expressed by murine monocytes or macrophages. CD4 increases the affinity in the interaction between T cell receptor (TCR) and major histocompatibility complex II (MHC II) antigen complex by binding to a non-polymorphic region of MHC II and acting as a co-receptor to TCR in MHC II-restricted antigen recognition. CD4 also amplifies signals from TCR to the cytoplasm through the interaction of its intracellular domain with the cytoplasmic tyrosine kinases such as lymphocyte-specific protein tyrosine kinase (Lck). This monoclonal antibody is widely used as a phenotypic marker for CD4 expression. The RM4-5 clone can block binding of the GK1.5 antibody clone to CD4 on the cell surface. The GK1.5 antibody clone is also reported to be cross-reactive with Syrian hamster CD4.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: PE
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Hamster;Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR