Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (Alexa Fluor® 488)

Lieferant: STEMCELL Technologies
60005AD
STMC60005ADEA 533 EUR
STMC60005AD
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (Alexa Fluor® 488)
Antikörper
Mouse monoclonal IgG1 antibody against human, chimpanzee CD19, Alexa Fluor® 488-conjugated.

  • B cells

The HIB19 antibody reacts with CD19, an ~95 kDa type 1 transmembrane glycoprotein expressed on the surface of B cells throughout all stages of development, from early pre-B cells to plasma cells. Expression is down-regulated but persists in terminally differentiated plasma cells. CD19 is also found on follicular dendritic cells. By associating with CD21 and CD81, CD19 functions as a co-receptor for the B cell receptor and is involved in B cell activation and differentiation. Activation of CD19 is accompanied by phosphorylation of the cytoplasmic domain, which promotes binding to kinases and the induction of intracellular signaling cascades. Mutations in CD19 can result in severe immunodeficiency syndromes. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human CD19 Positive Selection Kit (Catalog #18054), EasySep™ Human Whole Blood CD19 Positive Selection Kit (Catalog #18084), EasySep™ HLA Whole Blood B Cell Positive Selection Kit (Catalog #18184HLA); partial blocking may be observed, as well as EasySep™ HLA B Cell Enrichment: Complete Processing Kit for Whole Blood (Catalog #19954HLA) and EasySep™ HLA Total Lymphocyte Enrichment: Complete Processing Kit for Whole Blood (Catalog #19961HLA).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR