Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8] (Pacific Blue™)

Lieferant: STEMCELL Technologies
60031PB 60031PB.1
STMC60031PBEA 563 EUR
STMC60031PB STMC60031PB.1
Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8] (Pacific Blue™)
Antikörper
Rat monoclonal IgG2a antibody against mouse Ly-6G, Pacific Blue™-conjugated.

  • Granulocytes
  • Subsets
  • Myeloid cells

The 1A8 antibody reacts with Ly-6G, a 21 to 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen (Gr-1). Gr-1 is expressed on monocytes, neutrophils and subsets of macrophages, plasmacytoid dendritic cells, and T cells. Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation. In the periphery, Gr-1 is found predominantly on neutrophils and is a useful marker for these cells. The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C. It has been reported that the 1A8 antibody detects Ly-6G-expressing granulocytes in peripheral blood, whereas the RB6-8C5 antibody also binds to Ly-6C-expressing lymphocytes, monocytes, and dendritic cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ mouse Neutrophil enrichment kit (Catalog #19762).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-Ly-6G Rat Monoclonal Antibody [clone: 1A8] (Pacific Blue™)
Antigen: Ly-6G
Clonality: Monoclonal
Clone: 1A8
Conjugation: Pacific Blue™
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR