Anti-Oligodendrocyte Marker O4 Mouse Monoclonal Antibody [clone: 81]

Lieferant: STEMCELL Technologies
60053
STMC60053EA 1040 EUR
STMC60053
Anti-Oligodendrocyte Marker O4 Mouse Monoclonal Antibody [clone: 81]
Antikörper
Mouse monoclonal IgM antibody against human, mouse, rat oligodendrocyte marker O4, unconjugated.

  • Neural cells Psc Derived
  • Neural stem
  • Progenitor cells

The 81 antibody clone reacts with the O4 antigen, a sulfated galactocerebroside (sulfatide). O-Antigens are primarily expressed on the extracellular leaflet of the myelin plasma membrane of differentiating and mature oligodendrocytes in the central nervous system, as well as on Schwann cells in the peripheral nervous system. O4 is formed during postnatal development and is a marker for cell bodies and processes of oligodendrocytes types I and II. O4 is expressed in pro-oligodendrocytes, however, it is not found on O-2A-progenitor cells. Abnormal expression of O4 has been associated with various disorders, including neurofibromatosis and schizophrenia.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-Oligodendrocyte Marker O4 Mouse Monoclonal Antibody [clone: 81]
Antigen: Oligodendrocyte Marker O4
Clonality: Monoclonal
Clone: 81
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Chicken, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR