Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (PE)

Lieferant: STEMCELL Technologies
60062PE 60062PE.1
STMC60062PEEA 533 EUR
STMC60062PE STMC60062PE.1
Anti-SSEA-4 Mouse Monoclonal Antibody [clone: MC-813-70] (PE)
Antikörper
Mouse monoclonal IgG3 antibody against human, mouse, rat SSEA-4, PE-conjugated.

  • Pluripotent stem cells

The MC-813-70 antibody reacts with stage-specific embryonic antigen-4 (SSEA-4), a glycolipid carbohydrate antigen expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), undifferentiated embryonic stem (ES) and induced pluripotent stem (iPS) cells, a subset of mesenchymal stem cells, and rhesus monkey ES cell lines. No immunoreactivity is evident with undifferentiated mouse EC, EG, ES, and iPS cells. Expression of SSEA-4 is down-regulated following differentiation of human EC, ES, and iPS cells. In contrast, the differentiation of mouse EC, ES, or iPS cells may be accompanied by an increase in SSEA-4 expression. This antibody clone has been verified for labeling human ES and iPS cells grown in mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860), and has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Human ES/iPS Cell TRA-1-60 Positive Selection Kit (Catalog #18166) and EasySep™ hESC/hiPSC SSEA-4 Positive Selection Kit (Catalog #18165; partial blocking may be observed).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: SSEA-4
Clonality: Monoclonal
Clone: MC-813-70
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Cat, Chicken, Dog, Human, Mouse, Rabbit, Rat, Rhesus monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR