Mouse monoclonal IgM antibody against human, rat, rhesus TRA-1-81, APC-conjugated.
This monoclonal antibody reacts with a carbohydrate epitope on TRA-1-81, a high-molecular-mass (> 200 kDa) glycoprotein expressed on the surface of human embryonal carcinoma (EC), embryonic germ (EG), embryonic stem (ES), and induced pluripotent stem (iPS) cells, as well as rhesus monkey and rat ES cell lines. No immunoreactivity is observed with mouse EC, EG, ES, or iPS cells. The epitope of the TRA-1-81 antibody is associated with a keratan-sulfated transmembrane protein identified as podocalyxin and is resistant to treatment with neuraminidase.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Type: Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81] (APC)
Antigen: TRA-1-81
Clonality: Monoclonal
Clone: TRA-1-81
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Rat, Rhesus monkey