Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21]

Lieferant: STEMCELL Technologies
60070 60070.1
STMC60070EA 265 EUR
STMC60070 STMC60070.1
Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21]
Antikörper
Mouse monoclonal IgG1, kappa isotype control antibody, unconjugated.

The MOPC-21 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components. This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining).

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-Igg1 Isotype Control Mouse Monoclonal Antibody [clone: MOPC-21]
Antigen: Igg1 Isotype Control
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR