Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (Biotin)
Lieferant: STEMCELL Technologies
60109BT 60109BT.1
STMC60109BTEA 540 EUR
STMC60109BT STMC60109BT.1
Anti-CD62L Rat Monoclonal Antibody [clone: MEL-14] (Biotin)
Antikörper
Rat monoclonal IgG2a antibody against mouse CD62L (L-selectin), biotin-conjugated.
  • T cells
The MEL-14 antibody reacts with murine CD62L (L-Selectin or LECAM-1), an ~74 - 95 kDa type I glycoprotein expressed on the surface of a majority of leukocytes. CD62L mediates leukocyte-endothelial cell interactions through its association with ligands such as CD34, glyCAM-1, MAdCAM-1 and PSGL-1. It functions to facilitate lymphocyte rolling on the vascular endothelium, trafficking of lymphocytes to the lymph nodes, and homing of lymphocytes and neutrophils toMore Product Information