Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (APC)

Lieferant: STEMCELL Technologies
60110AZ 60110AZ.1
STMC60110AZEA 586 EUR
STMC60110AZ STMC60110AZ.1
Anti-CD123 (IL-3Rα) Mouse Monoclonal Antibody [clone: 6H6] (APC)
Antikörper
Mouse monoclonal IgG1 antibody against human, rhesus, sooty mangabey CD123 (IL-3Rα), APC-conjugated.

  • Dendritic cells
  • Granulocytes
  • Subsets
  • Hematopoietic Stem
  • Progenitor cells
  • Myeloid cells

The 6H6 antibody reacts with human CD123 (IL-3 receptor subunit α), an ~70 kDa type I transmembrane glycoprotein belonging to the type I cytokine receptor family (type 5 subfamily) and the immunoglobulin (Ig) superfamily. CD123 constitutes the ligand-binding α chain of the heterodimeric IL-3 receptor. CD123 binds IL-3 with low affinity per se, but when associated with CD131 (the signal-transducing β chain of the receptor), high-affinity binding of IL-3 is observed. CD123 is expressed by hematopoietic progenitor cells, endothelial cells, basophils, eosinophils, mast cells, monocytes, macrophages, dendritic cells, megakaryocytes, a subset of B cells, and by neutrophils if cultured in the presence of granulocyte-macrophage colony-stimulating factor (GM-CSF). IL-3 binding to CD123 stimulates proliferation, differentiation, and viability of hematopoietic cells. CD123 is highly expressed in malignancies such as acute leukemia. The 6H6 antibody does not inhibit binding of IL-3 to either CD123 or the IL-3 receptor.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD123 (IL-3Rα)
Clonality: Monoclonal
Clone: 6H6
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Rhesus monkey;Sooty mangabey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR