Anti-Red Blood Cell Mouse Monoclonal Antibody [clone: T3G6]

Lieferant: STEMCELL Technologies
60132
STMC60132EA 403 EUR
STMC60132
Anti-Red Blood Cell Mouse Monoclonal Antibody [clone: T3G6]
Antikörper
Mouse monoclonal IgG1 antibody against rhesus red blood cells, unconjugated.

  • Erythroid cells
  • Red blood cells

The mouse monoclonal antibody T3G6 recognizes mature red blood cells and erythroid precursor cells in rhesus monkey bone marrow. Antibody clone T3G6 has been developed in mice immunized with rhesus monkey (Macaca mulatta) red blood cells. The antigen to which clone T3G6 binds has not been identified. The reactivity with red blood cells from other monkey species has not been determined. Clone T3G6 does not recognize human red blood cells.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Red Blood Cell
Clonality: Monoclonal
Clone: T3G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Non-Human Primate
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR