Anti-Biotin Mouse Monoclonal Antibody [clone: C6D5.1.1]

Lieferant: STEMCELL Technologies
60145
STMC60145EA 688 EUR
STMC60145
Anti-Biotin Mouse Monoclonal Antibody [clone: C6D5.1.1]
Antikörper
Mouse monoclonal IgG1 antibody against biotin, unconjugated.

The C6D5.1.1 antibody reacts with biotin (vitamin H, vitamin B7, coenzyme R), a 244.3 Da molecule that is present in all living cells and is essential for cell growth and the citric acid cycle. Biotin is commonly coupled to proteins and other molecules without altering their biological activity for use in a number of applications. Both avidin and streptavidin form strong non-covalent bonds with biotin in a 1:4 ratio and thus efficiently cross-link biotinylated molecules. Both avidin and streptavidin can, however, exhibit nonspecific binding that can be problematic in certain applications. Cross-linking of biotinylated molecules, such as immunoglobulins, can also be achieved using anti-biotin antibodies such as clone C6D5.1.1. This approach may be preferable to eliminate the non-specific binding problems associated with streptavidin, or when a lesser degree of cross-linking is required.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: Biotin
Clonality: Monoclonal
Clone: C6D5.1.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR