Anti-Epithelial Cell Mouse Monoclonal Antibody [clone: 5E11.3.1]

Lieferant: STEMCELL Technologies
60147
STMC60147EA 545 EUR
STMC60147
Anti-Epithelial Cell Mouse Monoclonal Antibody [clone: 5E11.3.1]
Antikörper
Mouse monoclonal IgG1 antibody against human epithelial cells, unconjugated.

  • Mammary cells

The 5E11.3.1 antibody reacts with an unidentified marker expressed on the surface of most luminal cells of normal human mammary epithelium. Weak labeling (or no labeling) is observed within the basal cell compartment of the mammary epithelium. The 5E11.3.1 antibody gives variable labeling on malignant breast epithelium. Positive labeling is observed on the following cell lines: MCF7, CAMA, T47D, BT-10, BT-20, COLO 205, and WiDr. Labeling is negative on human bone marrow cells, blood cells, lymphoid and mesenchymal tissues, mammary and marrow fibroblast cultures, and human embryonic stem cell line H9.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-Epithelial Cell Mouse Monoclonal Antibody [clone: 5E11.3.1]
Antigen: Epithelial Cell
Clonality: Monoclonal
Clone: 5E11.3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR