Anti-CD10 Mouse Monoclonal Antibody [clone: FR4D11]

Lieferant: STEMCELL Technologies
60149
STMC60149EA 0 EUR
STMC60149
Anti-CD10 Mouse Monoclonal Antibody [clone: FR4D11]
Antikörper
Mouse monoclonal IgG1 antibody against human CD10, unconjugated.

  • Mammary cells

The FR4D11 antibody reacts with CD10, a 100 kDa type II transmembrane glycoprotein. CD10 is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt, and follicular germinal center lymphomas, immature and transitional B cells within adult bone marrow, and on blast cells from patients with chronic myelogenous leukemia (CML). CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts on the brush border of kidney, and gut epithelial cells. Most notably, CD10 was reported to mark human mammary bipotent progenitor cells.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD10
Clonality: Monoclonal
Clone: FR4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR