Anti-p63 (deltaN) Rabbit Polyclonal Antibody

Lieferant: STEMCELL Technologies
60154
STMC60154EA 795 EUR
STMC60154
Anti-p63 (deltaN) Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody against human p63 (deltaN), unconjugated.

  • Airway cells
  • Dermal cells

The Poly6190 antibody reacts with the deltaN isoforms of human p63, a member of the p53 family of transcription factors. Alternative splicing of the p63 transcript gives rise to two classes of isoforms. The TA isoforms (TA p63-α, -β, -γ) possess an N-terminal transactivation domain, while the deltaN isoforms (deltaN p63-α, -β, -γ) lack the transactivation domain. The TA isoforms are capable of transactivating p53 targets, whereas they antagonize p53. The deltaN isoforms are generally dominant to the TA isoforms and repress the latter. The p63 protein is localized to the nucleus and plays an important role in epithelial stem cell regeneration, epidermal morphogenesis, limb development, and stratification.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: p63 (deltaN)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR