Anti-MUC1 (CD227) Mouse Monoclonal Antibody [clone: 16A] (PE)

Lieferant: STEMCELL Technologies
60155PE 60155PE.1
STMC60155PEEA 0 EUR
STMC60155PE STMC60155PE.1
Anti-MUC1 (CD227) Mouse Monoclonal Antibody [clone: 16A] (PE)
Antikörper
Mouse monoclonal IgG1 antibody against human MUC1 (CD227), PE-conjugated.

  • Airway cells

The 16A antibody reacts with human MUC1, a large (>250 kDa) heavily glycosylated type 1 transmembrane protein expressed on the surface of most glandular and ductal epithelial cells and a variety of hematopoietic cells. A characteristic feature of the MUC1 glycoprotein is a core domain composed of a variable number of tandem repeats and multiple oligosaccharide side chains. Because the extracellular portion of MUC1 can extend beyond most cell surface proteins, it is thought to play a role in cell-cell and cell-substrate adhesion. The protein is highly expressed by a majority of human adenocarcinomas and is associated with a poor prognosis. In the mammary gland, MUC1 is localized on the apical plasma membrane of luminal epithelial cells. The clone 16A antibody has a higher affinity for the glycosylated form of MUC1.

Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: MUC1 (CD227)
Clonality: Monoclonal
Clone: 16A
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR